Anti-HPV33 E6 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-2968R-CY5
BOSSBS-2968R-CY5EA 1200 CHF
BOSSBS-2968R-CY5
Anti-HPV33 E6 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Human papilloma viruses (HPVs) can be classified as either high risk or low risk according to their association with cancer. HPV16 and HPV18 are the most common of the high risk group while HPV6 and HPV11 are among the low risk types. Approximately 90% of cervical cancers contain HPV DNA of the high risk types. Mutational analysis have shown that the E6 and E7 genes of the high risk HPVs are necessary and sufficient for HPV transforming function. The specific interactions of the E6 and E7 proteins with p53 and pRB, respectively, correlate with HPV high and low risk classifications. The highMore Product Information