Anti-CD44 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-2780R-A350
BOSSBS-2780R-A350EA 1190 CHF
BOSSBS-2780R-A350
Anti-CD44 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Receptor for hyaluronic acid (HA). Mediates cell-cell and cell-matrix interactions through its affinity for HA, and possibly also through its affinity for other ligands such as osteopontin, collagens, and matrix metalloproteinases (MMPs). Adhesion with HA plays an important role in cell migration, tumor growth and progression. In cancer cells, may play an important role in invadopodia formation. Also involved in lymphocyte activation, recirculation and homing, and in hematopoiesis. Altered expression or dysfunction causes numerous pathogenic phenotypes. Great protein heterogeneity due toMore Product Information