Anti-ADK Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-2778R
BOSSBS-2778REA 965 CHF
BOSSBS-2778R
Anti-ADK Rabbit Polyclonal Antibody
Antikörper
widespread effects on the cardiovascular, nervous, respiratory, and immune systems and inhibitors of ADK could play an important pharmacological role in increasing intravascular adenosine concentrations and acting as antiinflammatory agents. The encoded protein does not present any sequence similarities to other well characterized mammalian nucleoside kinases. In contrast, 2 regions were identified with significant sequence identity to microbial ribokinase and fructokinases and a bacterial inosine/guanosine kinase. Thus, ADK is a structurally distinct mammalian nucleoside kinase that appearsMore Product Information