Anti-SPHK1 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-2652R-CY7
BOSSBS-2652R-CY7EA 1210 CHF
BOSSBS-2652R-CY7
Anti-SPHK1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Sphingosine-1-phosphate (SPP) is a novel lipid messenger with both intracellular and extracellular functions. Intracellularly, it regulates proliferation and survival, and extracellularly, it is a ligand for EDG1 (MIM 601974). Various stimuli increase cellular levels of SPP by activation of sphingosine kinase (SPHK), the enzyme that catalyzes the phosphorylation of sphingosine. Competitive inhibitors of SPHK block formation of SPP and selectively inhibit cellular proliferation induced by a variety of factors, including platelet-derived growth factor (e.g., MIM 173430) and serum.[supplied byMore Product Information