Anti-CYP7A1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-2399R-A680
BOSSBS-2399R-A680EA 1200 CHF
BOSSBS-2399R-A680
Anti-CYP7A1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Catalyses a rate-limiting step in cholesterol catabolism and bile acid biosynthesis by introducing a hydrophilic moiety at position 7 of cholesterol. Important for cholesterol homeostasis.

Type: Primary
Antigen: CYP7A1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 351-400/504
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR