Anti-LGB Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-2065R-CY7
BOSSBS-2065R-CY7EA 1210 CHF
BOSSBS-2065R-CY7
Anti-LGB Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Bovine beta-lactoglobulin is the content of the milk in the category of a protein myosin. Protein contained in whole milk (3.7 percent) to 80% of the casein in the removal of the casein whey protein, 55% of the beta-lactoglobulin. Access to the crystallization of the isoelectric point = 5.2, the pH in this dual capacity under the main structure. The people in latex-free Beta-lactoglobulin. The beta-lactoglobulin and bov beta-lactoglobulinA-based.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LGB
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR