Anti-PTGER3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-1876R-A555
BOSSBS-1876R-A555EA 1170 CHF
BOSSBS-1876R-A555
Anti-PTGER3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The protein encoded by this gene is a member of the G-protein coupled receptor family. This protein is one of four receptors identified for prostaglandin E2 (PGE2). This receptor may have many biological functions, which involve digestion, nervous system, kidney reabsorption, and uterine contraction activities. Studies of the mouse counterpart suggest that this receptor may also mediate adrenocorticotropic hormone response as well as fever generation in response to exogenous and endogenous stimuli. Multiple transcript variants encoding different isoforms have been found for this gene. [More Product Information