Anti-PEG3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-1870R-CY5.5
BOSSBS-1870R-CY5.5EA 1210 CHF
BOSSBS-1870R-CY5.5
Anti-PEG3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Induces apoptosis in cooperation with SIAH1A. Acts as a mediator between p53/TP53 and BAX in a neuronal death pathway that is activated by DNA damage. Acts synergistically with TRAF2 and inhibits TNF induced apoptosis through activation of NF-kappa-B (By similarity). Possesses a tumor suppressing activity in glioma cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PEG3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR