Anti-GDF1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-1794R-CY5
BOSSBS-1794R-CY5EA 1200 CHF
BOSSBS-1794R-CY5
Anti-GDF1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
This gene encodes a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site that is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. This protein is involved in the establishment of left-right asymmetry in early embryogenesis and in neural development in later embryogenesis. This protein is transcribed from a monocistronic mRNA early inMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol GDF1
Environmentally Preferable
Antigen-Name Growth Differentiation Factor 1
Antigen-Synonyme DORV|RAI|DTGA3
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy5®
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Isotyp IgG
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR