Anti-GDF1 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-1794R-CY3
BOSSBS-1794R-CY3EA 1200 CHF
BOSSBS-1794R-CY3
Anti-GDF1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
This gene encodes a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site that is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. This protein is involved in the establishment of left-right asymmetry in early embryogenesis and in neural development in later embryogenesis. This protein is transcribed from a monocistronic mRNA early inMore Product Information