Anti-DRD4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-1746R-A647
BOSSBS-1746R-A647EA 1190 CHF
BOSSBS-1746R-A647
Anti-DRD4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
This gene encodes the D4 subtype of the dopamine receptor. The D4 subtype is a G-protein coupled receptor which inhibits adenylyl cyclase. It is a target for drugs which treat schizophrenia and Parkinson disease. Mutations in this gene have been associated with various behavioral phenotypes, including autonomic nervous system dysfunction, attention deficit/hyperactivity disorder, and the personality trait of novelty seeking. This gene contains a polymorphic number (2-10 copies) of tandem 48 nt repeats; the sequence shown contains four repeats.DRD4 expression has been reported in variousMore Product Information