Anti-DUT Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-1721R-CY7
BOSSBS-1721R-CY7EA 1200 CHF
BOSSBS-1721R-CY7
Anti-DUT Rabbit Polyclonal Antibody (Cy7®)
Antikörper
This enzyme is involved in nucleotide metabolism: it produces dUMP, the immediate precursor of thymidine nucleotides and it decreases the intracellular concentration of dUTP so that uracil cannot be incorporated into DNA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DUT
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR