Anti-AGT Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-1707R-CY3
BOSSBS-1707R-CY3EA 1200 CHF
BOSSBS-1707R-CY3
Anti-AGT Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The protein encoded by this gene, pre-angiotensinogen or angiotensinogen precursor, is expressed in the liver and is cleaved by the enzyme renin in response to lowered blood pressure. The resulting product, angiotensin I is then cleaved by angiotensin converting enzyme (ACE) to generate the physiologically active enzyme angiotensin II. The protein is involved in maintaining blood pressure and in the pathogenesis of essential hypertension and preeclampsia. active peptide sequence:DRVYIHPFHL

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AGT
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR