Anti-TNFRSF10B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-1696R-HRP
BOSSBS-1696R-HRPEA 1180 CHF
BOSSBS-1696R-HRP
Anti-TNFRSF10B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Receptor for the cytotoxic ligand TNFSF10/TRAIL. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. Promotes the activation of NF-kappa-B. Essential for ER stress-induced apoptosis (By similarity).

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: TNFRSF10B
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR