Anti-HBsAg Goat polyclonal antibody

Lieferant: Bioss
BS-1557G
BOSSBS-1557GEA 938 CHF
BOSSBS-1557G
Anti-HBsAg Goat polyclonal antibody
Antikörper
Type: Primary
Antigen: HBsAg
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR