Anti-IDUA Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)

Lieferant: Bioss
BS-15542R-A555
BOSSBS-15542R-A555EA 1190 CHF
BOSSBS-15542R-A555
Anti-IDUA Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)
Antikörper
This gene encodes an enzyme that hydrolyses the terminal alpha-L-iduronic acid residues of two glycosaminoglycans, dermatan sulfate and heparan sulfate. This hydrolysis is required for the lysosomal degradation of these glycosaminoglycans. Mutations in this gene that result in enzymatic deficiency lead to the autosomal recessive disease mucopolysaccharidosis type I (MPS I).

Type: Primary
Antigen: IDUA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 101-200/653
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR