Anti-C8ORF76 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
Lieferant: Bioss
BS-15301R-A350
BOSSBS-15301R-A350EA 1190 CHF
BOSSBS-15301R-A350
Anti-C8ORF76 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
Antikörper
C8orf76 (chromosome 8 open reading frame 76) is a 380 amino acid protein encoded by a gene that maps to human chromosome 8q24.13. Consisting of nearly 146 million bases, chromosome 8 encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and are typically associated with a poor prognosis. Portions of chromosome 8 have been linked to schizophrenia and bipolar disorder. Trisomy 8, also known as Warkany syndrome 2, most often results in early miscarriage but is occasionally seen in a mosaic form inMore Product Information