Anti-C3orf55 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-15178R
BOSSBS-15178REA 965 CHF
BOSSBS-15178R
Anti-C3orf55 Rabbit Polyclonal Antibody
Antikörper
Chromosome 3 is made up of about 214 million bases encoding over 1,100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumor suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesis suppressor SEMA3B. Marfan Syndrome, porphyria, von Hippel-Lindau syndrome, osteogenesis imperfecta and Charcot-Marie-Tooth Disease are a few of the numerous genetic diseases associated withMore Product Information