Anti-C20orf14 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-15094R-HRP
BOSSBS-15094R-HRPEA 1200 CHF
BOSSBS-15094R-HRP
Anti-C20orf14 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The protein encoded by this gene appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. The encoded protein also can bind androgen receptor, providing a link between transcriptional activation and splicing.

Type: Primary
Antigen: C20orf14
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 331-430/941
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR