Anti-C20orf14 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-15094R-A750
BOSSBS-15094R-A750EA 1190 CHF
BOSSBS-15094R-A750
Anti-C20orf14 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The protein encoded by this gene appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. The encoded protein also can bind androgen receptor, providing a link between transcriptional activation and splicing.

Type: Primary
Antigen: C20orf14
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 331-430/941
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

TECHNISCHE DATEN

Antigen-Name C20orf14
Environmentally Preferable
Antigen-Synonyme wu:fa05f07|PRP6_HUMAN|wu:fc12b02|PRPF6|U5 snRNP associated 102 kDa protein|Pre mRNA processing factor 6|PRPF 6|fc12b02|zgc:65913|Chromosome 20 open reading frame 14|Putative mitochondrial outer membrane protein import receptor|Androgen receptor N terminal domain transactivating protein 1|hypothetical protein LOC323855|U5 102 kDa protein|PRP6 homolog|p102 U5 small nuclear ribonucleoprotein particle binding protein
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 750
Reaktivität
Wirt Rabbit
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Modification Unmodified
Epitop 331-430/941
Immunogen KLH conjugated synthetic peptide derived from human C20orf14
Form Liquid
Molecular weight 107 kDa
Reinigung Purified by Protein A
Konzentration 1 µg/µl
Lagerungspuffer Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
Lagertemperatur Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transporttemperatur 4 °C
Validierte Anwendungen ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR