Anti-C20orf14 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-15094R-A350
BOSSBS-15094R-A350EA 1190 CHF
BOSSBS-15094R-A350
Anti-C20orf14 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The protein encoded by this gene appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. The encoded protein also can bind androgen receptor, providing a link between transcriptional activation and splicing.

Type: Primary
Antigen: C20orf14
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 331-430/941
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR