Anti-C1orf64 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-15073R-FITC
BOSSBS-15073R-FITCEA 1200 CHF
BOSSBS-15073R-FITC
Anti-C1orf64 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. TheMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Name C1orf64
Environmentally Preferable
Antigen-Synonyme CA064_HUMAN|C1orf64|MGC24047|RP11-5P18.4|Uncharacterized protein C1orf64
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation FITC (Fluorescein Isothiocyanate)
Reaktivität Human
Wirt Rabbit
Gen-ID 149563
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Modification Unmodified
Epitop 81-169/169
Immunogen KLH conjugated synthetic peptide derived from human C1orf64
Form Liquid
Molecular weight 18 kDa
Reinigung Purified by Protein A
Konzentration 1 µg/µl
Lagerungspuffer Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
Lagertemperatur Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transporttemperatur 4 °C
Validierte Anwendungen ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR