Anti-C1orf55 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-15071R
BOSSBS-15071REA 965 CHF
BOSSBS-15071R
Anti-C1orf55 Rabbit Polyclonal Antibody
Antikörper
Belonging to the UPF0667 family, C1orf55 is a 451 amino acid protein that is phosphorylated upon DNA damage, likely by Atm or ATR. There are three different isoforms of C1orf55 that are produced as a result of alternative splicing events. The gene encoding C1orf55 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging diseaseMore Product Information