Anti-C1orf104 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-15002R-A555
BOSSBS-15002R-A555EA 1190 CHF
BOSSBS-15002R-A555
Anti-C1orf104 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. TheMore Product Information