Anti-C17orf87 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-13685R
BOSSBS-13685REA 965 CHF
BOSSBS-13685R
Anti-C17orf87 Rabbit Polyclonal Antibody
Antikörper
C17orf87 (chromosome 17 open reading frame 87) is a 145 amino acid protein that is encoded by a gene mapping to human chromosome 17. Chromosome 17 makes up over 2.5% of the human genome with about 81 million bases encoding over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and Li-Fraumeni syndrome. Like p53, BRCA1 isMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol C17orf87
Environmentally Preferable
Antigen-Name Chromosome 17 open reading frame 87
Antigen-Synonyme SCIMP|SLP65/SLP76|DTFT5783|Transmembrane protein C17orf87|SLP adaptor and CSK interacting membrane protein|UNQ5783/PRO16090.|CQ087_HUMAN|UNQ5783|C17orf87
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A.
Konzentration 1 ug/ul
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Transporttemperatur 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR