Anti-TMEM176A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-13623R-A647
BOSSBS-13623R-A647EA 1190 CHF
BOSSBS-13623R-A647
Anti-TMEM176A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
TMEM176A is a 235 amino acid multi-pass membrane protein belonging to the TMEM176 family. The gene encoding GS188 maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Chromosome 7 has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterized by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance. Deletions ofMore Product Information