Anti-GBE1 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-13300R-CY3
BOSSBS-13300R-CY3EA 1190 CHF
BOSSBS-13300R-CY3
Anti-GBE1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
GBE1 is a 702 amino acid protein that is expressed at high levels in muscle and liver and is involved in glycogen biosynthesis. Existing as a monomer, GBE1 catalyzes the transfer of alpha-1,4-linked glucosyl units from the outer end of a glycogen chain to an alpha-1,6 position on a neighboring glycogen chain and, via this catalytic activity, plays an essential role in glycogen accumulation. Defects in the gene encoding GBE1 are the cause of glycogen storage disease type 4 (GSD4) and adult polyglucosan body disease (APBD), the first of which is a metabolic disorder that is associated with theMore Product Information