Anti-GALNT11 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-13271R-A647
BOSSBS-13271R-A647EA 1190 CHF
BOSSBS-13271R-A647
Anti-GALNT11 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
The UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes are substrate-specific proteins that catalyze the transfer of GalNAc (N-acetylgalactosamine) to serine and threonine residues onto various proteins, thereby initiating mucin-type O-linked glycosylation in the Golgi apparatus. GalNAc-T11 (Polypeptide N-acetylgalactosaminyltransferase 11), also known as UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 11, is a 608 amino acid protein that catalyzes glycosylation of Muc1, Muc4.1 and EA2, though it does not display enzymaticMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol GALNT11
Environmentally Preferable
Antigen-Name Polypeptide N-Acetylgalactosaminyltransferase 11
Antigen-Synonyme GALNACT11|GALNAC-T11
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 647
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 63917
Isotyp IgG
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR