Anti-GALNS Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-13269R-A350
BOSSBS-13269R-A350EA 1190 CHF
BOSSBS-13269R-A350
Anti-GALNS Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Chondroitinase is a 522 amino acid protein that localizes to the lysosome and functions as an exohydrolase that is essential for the degradation of glycosaminoglycans, keratan sulfate and chondroitin 6-sulfate. Using calcium as a cofactor, Chondroitinase, which exists as a disulfide linked oligomer, catalyzes the hydrolysis of the 6-sulfate group on target substrates. Defects in the gene encoding Chondroitinase are the cause of mucopolysaccharidosis type 4A (MPS4A), an autosomal recessive lysosomal storage disease that is characterized by the intracellular accumulation of keratan sulfate andMore Product Information