Anti-FLNA Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-13164R
BOSSBS-13164REA 989 CHF
BOSSBS-13164R
Anti-FLNA Rabbit Polyclonal Antibody
Antikörper
Promotes orthogonal branching of actin filaments and links actin filaments to membrane glycoproteins. Anchors various transmembrane proteins to the actin cytoskeleton and serves as a scaffold for a wide range of cytoplasmic signaling proteins. Interaction with FLNA may allow neuroblast migration from the ventricular zone into the cortical plate. Tethers cell surface-localized furin, modulates its rate of internalization and directs its intracellular trafficking.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Filamin A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
More Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol FLNA
Antigen-Name Filamin A, alpha
Environmentally Preferable
Antigen-Synonyme OPD1|OPD2|XLVD|FMD|OPD|CVD1|XMVD|FLN-A|FLN|FLN1|ABP-280|NHBP|MNS|ABPX|CSBS
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 2316
Isotyp IgG
ImmunoChemie Yes
Modification Ser1084
Form liquid
Reinigung Purified by Protein A.
Konzentration 1 ug/ul
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Transporttemperatur 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR