Anti-EHM2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-13065R-HRP
BOSSBS-13065R-HRPEA 1200 CHF
BOSSBS-13065R-HRP
Anti-EHM2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
EHM2 belongs to the NF2/ERM/4.1 superfamily of proteins, which function in connecting cell surface transmembrane proteins to cytoskeletal molecules.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: EHM2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR