Anti-CTBS Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-12937R-CY5
BOSSBS-12937R-CY5EA 1200 CHF
BOSSBS-12937R-CY5
Anti-CTBS Rabbit Polyclonal Antibody (Cy5®)
Antikörper
CTBS is an evolutionarily conserved member of the glycosyl hydrolase 18 family of proteins. Localizing to the lysosome, CTBS plays a role in the degradation of asparagine-linked (Asn-linked) glycoproteins. Glycoproteins are translocated to lysosomes via endocytosis or autophagy where they are broken down by proteases and glycosidases. The catabolism of glycoproteins is an important step in the regular turnover of cellular contents and in maintaining the homeostasis of glycosylation. CTBS functions as a glycosidase that cleaves the reducing end GlcNAc from the core chitobiase unit ofMore Product Information