Anti-ATF2 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-12536R
BOSSBS-12536REA 989 CHF
BOSSBS-12536R
Anti-ATF2 Rabbit Polyclonal Antibody
Antikörper
ATF2 is a member of the ATF/CREB family of basic region leucine zipper DNA binding proteins that regulates transcription by binding to a consensus cAMP response element (CRE) in the promoter of various viral and cellular genes. Many of these genes are important in cell growth and differentiation, and in stress and immune responses. ATF2 is a nuclear protein that binds DNA as a dimer and can form dimers with members of the ATF/CREB and Jun/Fos families. It is a stronger activator as a heterodimer with cJun than as a homodimer. Several isoforms of ATF2 arise by differential splicing. TheMore Product Information