Anti-ADH6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-12449R-FITC
BOSSBS-12449R-FITCEA 1200 CHF
BOSSBS-12449R-FITC
Anti-ADH6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
ADH6 (alcohol dehydrogenase 6), also known as ADH-5, is a 368 amino acid member of the class V zinc-containing alcohol dehydrogenase family. This family of enzymes functions to metabolize a wide variety of substrates such as retinol, hydroxysteroids, ethanol, aliphatic alcohols and lipid peroxidation products. Localized to the cytoplasm and expressed in the stomach and liver, ADH6 catalyzes the reversible oxidation of alcohols to their corresponding aldehydes or ketones and is able to bind two zinc ions as cofactors. ADH6 contains a glucocorticoid response element upstream of its 5' UTRMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol ADH6
Environmentally Preferable
Antigen-Name Alcohol Dehydrogenase 6
Antigen-Synonyme ADH-5
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation FITC (Fluorescein Isothiocyanate)
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 130
Isotyp IgG
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR