Anti-RDH10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-11941R-A750
BOSSBS-11941R-A750EA 1190 CHF
BOSSBS-11941R-A750
Anti-RDH10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Retinol dehydrogenase with a clear preference for NADP. Converts all-trans-retinol to all-trans-retinal. Has no detectable activity towards 11-cis-retinol, 9-cis-retinol and 13-cis-retinol.

Type: Primary
Antigen: RDH10
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/341
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR