Anti-CBLN4 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11816R
BOSSBS-11816REA 965 CHF
BOSSBS-11816R
Anti-CBLN4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CBLN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR