Anti-SLCO3A1/OATP-D Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-11802R-A680
BOSSBS-11802R-A680EA 1190 CHF
BOSSBS-11802R-A680
Anti-SLCO3A1/OATP-D Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The organic anion transporter family of proteins mediate hepatic uptake of cardiac glycosides. OATP-D is a 710 amino acid member of the organic anion transporter protein family. As a multi-pass membrane protein, OATP-D mediates the Na+-independent transport of vasopressin, prostaglandins (PG) E1 and E2, thyroxine (T4), deltorphin II and other organic anions, but not estrone-3-sulfate, DPDPE, taurocholate, DHEAS or digoxin. OATP-D is ubiquitously expressed with highest levels present in leukocytes and spleen. OATP-D is expressed as four isoforms produced by alternative splicing events.