Anti-BLMH Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-11654R-A750
BOSSBS-11654R-A750EA 1190 CHF
BOSSBS-11654R-A750
Anti-BLMH Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Bleomycin hydrolase (BMH) is a cytoplasmic cysteine peptidase that is highly conserved through evolution; however, the only known activity of the enzyme is metabolic inactivation of the glycopeptide bleomycin (BLM), an essential component of combination chemotherapy regimens for cancer. The protein contains the signature active site residues of the cysteine protease papain superfamily.

Type: Primary
Antigen: BLMH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 308-400/455
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

TECHNISCHE DATEN

Environmentally Preferable
Antigen-Name BLMH
Antigen-Synonyme BH|BMH|BLMH_HUMAN|BLM hydrolase|EC 3.4.22.40|bleomycin hydrolase
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 750
Reaktivität Rat,Mouse
Wirt Rabbit
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Modification Unmodified
Epitop 308-400/455
Immunogen KLH conjugated synthetic peptide derived from human BLMH
Form Liquid
Molecular weight 53 kDa
Reinigung Purified by Protein A
Konzentration 1 µg/µl
Lagerungspuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagertemperatur Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transporttemperatur 4 °C
Validierte Anwendungen ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR