Anti-RTP3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-11619R-FITC
BOSSBS-11619R-FITCEA 1200 CHF
BOSSBS-11619R-FITC
Anti-RTP3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Members of the RTP (receptor transporter proteins) family have recently been discovered to influence bitter taste receptor expression in addition to inducing the expression of mammalian odorant receptors. RTP3 is a 232 amino acid single-pass type III membrane protein belonging to the TMEM7 family. Unlike other RTP proteins, RTP3 is not expressed in olfactory neurons but is expressed predominantly in liver. RTP3 is involved in the functional expression of bitter taste receptors and suppresses cell proliferation, and is also found in human circumvallate papillae and testis (regions whereMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol RTP3
Antigen-Name Receptor (chemosensory) Transporter Protein 3
Environmentally Preferable
Antigen-Synonyme RTP3_HUMAN|Receptor-transporting protein 3|LTM1|Receptor chemosensory transporter protein 3|TMEM7|RTP3|Receptor transporter protein 3|Transmembrane protein 7.
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation FITC (Fluorescein Isothiocyanate)
Reaktivität Human,Mouse
Wirt Rabbit
Isotyp IgG
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR