Anti-PCYT1A Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-11306R-CY7
BOSSBS-11306R-CY7EA 1200 CHF
BOSSBS-11306R-CY7
Anti-PCYT1A Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Increase in fetal surfactant synthesis and lung maturity is caused by the glucocorticoidal induction of enzymes required for phosphatidylcholine synthesis towards the end of gestation (1). The regulation of gestational age-dependent induction of phosphatidylcholine synthesis by glucocorticoids is still unclear (1). The rate-controlling enzyme in the phosphatidylcholine biosynthetic pathway is CTP-phosphocholine cytidylyltransferase A (CCT A) (2–4). In cultured eukaryotic cells, this enzyme is essential for survival (3). The alpha isoform is located in the nucleus and is regulated byMore Product Information