Anti-RNF60 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-11029R-CY5
BOSSBS-11029R-CY5EA 1200 CHF
BOSSBS-11029R-CY5
Anti-RNF60 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Midline-2 is a 715 amino acid protein encoded by the human gene MID2. Midline-2 belongs to the TRIM/RBCC family and contains two B box-type zinc fingers, one B30.2/SPRY domain, one COS domain, one fibronectin type-III domain and one RING-type zinc finger. Midline-2 is a cytoplasmic protein found as a homodimer or heterodimer with Midline-1. It also interacts with IGBP1 (Lymphocyte signaling protein A4). Dimerization is mediated by the tripartite motif (RBCC; RING- and B box-type zinc fingers and coiled coil domains) and microtubule association is dependent on the C-terminal B30.2 domain.More Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol RNF60
Antigen-Name Ring Finger Protein 60
Environmentally Preferable
Antigen-Synonyme Midline-2
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy5®
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 26040
Isotyp IgG
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR