Anti-ILVBL Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-11014R-CY5
BOSSBS-11014R-CY5EA 1200 CHF
BOSSBS-11014R-CY5
Anti-ILVBL Rabbit Polyclonal Antibody (Cy5®)
Antikörper
ILVBL is a 632 amino acid single-pass membrane protein that belongs to the TPP enzyme family. Expressed in the majority of tissues, ILVBL has the highest level of expression in heart, pancreas and placenta. ILVBL is highly homologous to several bacterial enzymes, including the B isozyme of the large catalytic subunit of E. coli acetohydroxy-acid synthase (AHAS) and the oxalyl-coA decarboxylase of O. formigenes, that utilize thiamine pyrophosphate as a cofactor. ILVBL binds one magnesium ion and one thiamine pyrophosphate per subunit, and may catalyze the initial step in branched-chain aminoMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol ILVBL
Antigen-Name IlvB (bacterial acetolactate synthase)-like
Environmentally Preferable
Antigen-Synonyme 209L8|ilvB-like protein|AHAS|acetolactate synthase-like protein|ILV2H|acetolactate synthase homolog
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy5®
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 10994
Isotyp IgG
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR