Anti-ILVBL Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-11014R-A750
BOSSBS-11014R-A750EA 1190 CHF
BOSSBS-11014R-A750
Anti-ILVBL Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
ILVBL is a 632 amino acid single-pass membrane protein that belongs to the TPP enzyme family. Expressed in the majority of tissues, ILVBL has the highest level of expression in heart, pancreas and placenta. ILVBL is highly homologous to several bacterial enzymes, including the B isozyme of the large catalytic subunit of E. coli acetohydroxy-acid synthase (AHAS) and the oxalyl-coA decarboxylase of O. formigenes, that utilize thiamine pyrophosphate as a cofactor. ILVBL binds one magnesium ion and one thiamine pyrophosphate per subunit, and may catalyse the initial step in branched-chain aminoMore Product Information
Order Now

TECHNISCHE DATEN

Environmentally Preferable
Antigen-Name ILVBL
Antigen-Synonyme IlvB-like protein|Acetolactate synthase homolog|Ilvbl|209L8|Acetolactate synthase-like protein|ILVBL_HUMAN|IlvB like protein|AHAS|IlvB bacterial acetolactate synthase like|ILV2H|Acetolactate synthase like protein
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 750
Reaktivität Human
Wirt Rabbit
Gen-ID 10994
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Durchflusszytometrie Yes
Modification Unmodified
Epitop 301-400/632
Immunogen KLH conjugated synthetic peptide derived from human ILVBL
Form Liquid
Molecular weight 68 kDa
Reinigung Purified by Protein A
Konzentration 1 µg/µl
Lagerungspuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagertemperatur Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transporttemperatur 4 °C
Validierte Anwendungen ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR