Anti-ILVBL Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-11014R-A555
BOSSBS-11014R-A555EA 1190 CHF
BOSSBS-11014R-A555
Anti-ILVBL Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
ILVBL is a 632 amino acid single-pass membrane protein that belongs to the TPP enzyme family. Expressed in the majority of tissues, ILVBL has the highest level of expression in heart, pancreas and placenta. ILVBL is highly homologous to several bacterial enzymes, including the B isozyme of the large catalytic subunit of E. coli acetohydroxy-acid synthase (AHAS) and the oxalyl-coA decarboxylase of O. formigenes, that utilize thiamine pyrophosphate as a cofactor. ILVBL binds one magnesium ion and one thiamine pyrophosphate per subunit, and may catalyze the initial step in branched-chain aminoMore Product Information