Anti-B3GALNT1 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-10438R
BOSSBS-10438REA 965 CHF
BOSSBS-10438R
Anti-B3GALNT1 Rabbit Polyclonal Antibody
Antikörper
This gene is a member of the beta-1,3-galactosyltransferase (beta3GalT) gene family. This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine). The beta3GalT genes are distantly related to the Drosophila Brainiac gene and have the protein coding sequence contained in a single exon. The beta3GalT proteins also contain conserved sequences not found in the beta4GalT or alpha3GalT proteins. TheMore Product Information