Anti-VAMP1 / VAMP2 / VAMP3 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-10427R
BOSSBS-10427REA 965 CHF
BOSSBS-10427R
Anti-VAMP1 / VAMP2 / VAMP3 Rabbit Polyclonal Antibody
Antikörper
Synaptobrevins/VAMPs, syntaxins, and the 25-kD synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. This gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Because of its high homology to other known VAMPs, its broad tissue distribution, and its subcellular localization, the protein encoded by this gene was shown to be the human equivalent of the rodent cellubrevin. In platelets the protein resides on a compartment that is not mobilized toMore Product Information