Anti-LAT Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-10126R-A750
BOSSBS-10126R-A750EA 1190 CHF
BOSSBS-10126R-A750
Anti-LAT Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Linker for activation of T cells (LAT) is an integral membrane protein (36 to 38 kD), which plays an important role in linking engagement of the TCR to the biochemical events of T cell activation. LAT antibody stains thymus and peripheral lymphoid tissues such as T cell areas in lymph nodes and spleen. LAT is expressed in T lymphocytes in interstitial spaces, platelets and megakaryocytes. LAT is not expressed in B cells, macrophages, plasma cells, monocytes, epithelial histiocytes and dendritic cells.

Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
PublicMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR