Anti-MUC13 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-10074R-CY7
BOSSBS-10074R-CY7EA 1200 CHF
BOSSBS-10074R-CY7
Anti-MUC13 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
MUC13 is a mucin. These are secreted or transmembrane glycoproteins that are expressed mainly in the digestive tract and on epithelial surfaces. MUC13 is upregulated in gastric cancer. Mucins have also been implicated in inflammatory bowel diseases.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: MUC13
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR