Anti-SNCA Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-0968R
BOSSBS-0968REA 965 CHF
BOSSBS-0968R
Anti-SNCA Rabbit Polyclonal Antibody
Antikörper
May be involved in the regulation of dopamine release and transport. Induces fibrillization of microtubule-associated protein tau. Reduces neuronal responsiveness to various apoptotic stimuli, leading to a decreased caspase-3 activation.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SNCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR