Anti-CD44 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-0521R-CY3
BOSSBS-0521R-CY3EA 1210 CHF
BOSSBS-0521R-CY3
Anti-CD44 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Receptor for hyaluronic acid (HA). Mediates cell-cell and cell-matrix interactions through its affinity for HA, and possibly also through its affinity for other ligands such as osteopontin, collagens, and matrix metalloproteinases (MMPs). Adhesion with HA plays an important role in cell migration, tumor growth and progression. In cancer cells, may play an important role in invadopodia formation. Also involved in lymphocyte activation, recirculation and homing, and in hematopoiesis. Altered expression or dysfunction causes numerous pathogenic phenotypes. Great protein heterogeneity due toMore Product Information